Find methods for your needs
Simple gradient method for the separation of estrogens and progesterones
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of strogens and progesterones. The separation is performed using a Thermo Scientific Acclaim Phenyl-1 column, with UV detection at 220 and 240 nm. for Research Use Only. Not for use in diagnostic procedures.| Market: | Clinical Research |
| Keywords: | 17-Ethynylestradiol, Acclaim Phenyl-1, Estradiol, Estriol, Estrogen, Estrone, HPLC, Norethindrone, Norgestrel, Progesterone, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.