Find methods for your needs
High resolution separation of Cytochrome C tryptic peptides
Description
The Thermo Scientific HPLC system is applied for the analysis of Cytochrome C tryptic peptides. The separation is performed using a Thermo Scientific Acclaim 300 C18 column, with UV detection at 214 nm.| Market: | BioPharma |
| Keywords: | Acclaim 300 C18, Bovine, Cytochrome C, HPLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.