Find methods for your needs
Rapid screening method for polycyclic aromatic hydrocarbons (PAHs) using an advanced solid core UHPLC column and system combination
Description
Demonstration of the advantages of using the Thermo Scientific Accucore Vanquish C18+, 1.5 μm column and Thermo Scientific Vanquish UHPLC system for the fast analysis of polycyclic aromatic hydrocarbons (PAHs). The advanced capabilities of the Vanquish UHPLC system allow the Accucore Vanquish columns to be operated at high flow rates that enable development of rapid analytical methods while maintaining high performance| Market: | Environmental |
| Keywords: | Polycyclic aromatic hydrocarbons (PAHs), UV detection, Vanquish UHPLC, Accucore Vanquish C18+ |
| Matrix: | N/A |
| Author: | Ken Meadows |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.