Find methods for your needs
Fast analysis of 1-naphthyl isothiocyanate (NITC) derivatives of volatile ethanolamines using normal-phase chromatography
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of 1-naphthyl isothiocyanate (NITC) derivatives of volatile ethanolamines. The separation is performed in normal-phase mode on a Thermo Scientific Acclaim Mixed-Mode HILIC-1 HPLC column, with UV detection at 280 nm.| Market: | Environmental |
| Keywords: | 1-naphthyl isothiocyanate, Acclaim Mixed-Mode HILIC-1, Diethylenetriamine-NITC, Ethylenediamine-NITC, HPLC, NITC, Normal-phase, Triethylenetetramine-NITC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.