Find methods for your needs
Fast analysis of 1-naphthyl isothiocyanate (NITC) derivatives of volatile alkylamines using normal-phase chromatography
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of 1-naphthyl isothiocyanate (NITC) derivatives of volatile alkylamines. The separation is performed in normal-phase mode on a Thermo Scientific Acclaim Mixed-Mode HILIC-1 HPLC column, with UV detection at 280 nm.| Market: | Environmental |
| Keywords: | Acclaim Mixed-Mode HILIC-1, Diethylamine-NITC, Dimethylamine-NITC, Ethylamine-NITC, HPLC, Methylamine-NITC, NITC, Normal-phase, UV detection, 1-naphthyl isothiocyanate |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.