Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of sorbitol in mouthwash. The separation is performed on a Thermo Scientific Accucore 150-Amide-HILIC HPLC column, with RI detection.Market: | Pharma |
Keywords: | Accucore 150-Amide-HILIC, HPLC, Mouthwash, RI detection, Sorbitol |
Matrix: | Mouthwash |
For Research Use Only. Not for use in diagnostic procedures.