Find methods for your needs
Comparison of the polyacrylic acid elution profile using size exclusion chromatography (SEC)
Description
The Thermo Scientific HPLC system is applied for the size exclusion chromatography analysis of polyacrylic acid. The separation is performed on a Thermo Scientific Acclaim SEC-300 and an Acclaim SEC-1000 HPLC column, with RI detection. The results are compared.| Market: | Environmental |
| Keywords: | Acclaim SEC-1000, Acclaim SEC-300, HPLC, Polyacrylic acid, Polymer, RI detection, SEC, Size exclusion chromatography |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.