Find methods for your needs
AN69: Determination of Aluminum in Complex Matrices Using Chelation Ion Chromatography
Description
Determination of trace metals in complex matrices has been a difficult analytical challenge. This application note describes the extension of Chelation ion chromatography for the analytical determination of aluminum in complex matrices. This technique allows aluminum to be preconcentrated from matrices high in salts, acids, or bases. Following selective concentration, aluminum is eluted to a conventional cation exchange column for separation from other transition metals. Selective detection is accomplished using post-column derivatization with Tiron and subsequent UV detection at 310 nm.| Market: | Environmental |
| Keywords: | Post column reaction, UV detection, waste water, Aluminum, Complex matrices, Transition metal, Chelation Ion Chromatography, Soils, Ground/surface water |
| Matrix: | Ground/Surface water, Sea water, Soils, Tissue/Biological samples, Waste water |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.