Find methods for your needs
Accurate determination of amlodipine and perindopril in solid dosage formulation using HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of amlodipine and perindopril in solid dosage formulation. The separation was performed on a Thermo Scientific Hypersil GOLD C8 HPLC column using a reversed-phase method with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Amlodipine, Formulation, HPLC, Hypersil GOLD C8, UV detection, Perindopril |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.