Find methods for your needs
Analysis of organic impurities of celecoxib using the method described in the USP Be the first to rate this application
Description
In this work the separation of celecoxib and impurities was performed using the Thermo Scientific BetaBasic Phenyl HPLC column according to the method described in the USP. As the temperature of the column needs to be set to 60 °C the Thermo Scientific Ultimate 3000 LC system was used with a pre-column heater to ensure the mobile phase was at the correct temperature. The resolution obtained between celecoxib and impurity A, and between celecoxib and impurity B met the USP requirements of greater than 1.8.
Market: Pharma
Keywords: HPLC, USP, UV detection, Impurities, Celecoxib, BetaBasic Phenyl
Matrix: N/A
Uploaded on 3/23/2015.
For Research Use Only. Not for use in diagnostic procedures.