Find methods for your needs
Carbohydrates in Dietary Fiber Using Capillary HPAE-IPAD Be the first to rate this application
Description
Cap_PA20_Dietary Fiber. This method shows the separation of expected carbohydrates in dietary fiber sample using a capillary CarboPac PA20 column using electrolytically-generated eluents and a capillary ICS-5000 IC system.
Market: Pharma
Matrix: Dietary fiber
Uploaded on 11/14/2010.
For Research Use Only. Not for use in diagnostic procedures.