Find methods for your needs
Simple HPLC-UV method for the determination of itraconazole impurities
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of itraconazole impurities. The separation was performed on a Thermo Scientific Accucore C18 HPLC column with UV detection at 225 nm.| Market: | Pharma |
| Keywords: | Accucore C18, HPLC, Itraconazole, UV detection, Impurities |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.