Find methods for your needs
Detection of Organochlorine Pesticides by GC-ECD Following U.S. EPA Method 8081
Description
The application describes a convenient method for analyzing and quantifying organochlorinated pesticides in extracts from solid and liquid matrices using a gas chromatograph (GC) equipped with an electron capture detector (ECD), according to the US regulation EPA 8081. The ECD detector has been demonstrated to be a sensitive, cheaper, and easier-to operate alternative to mass spectrometry, showing an excellent selectivity toward organohalogenated compounds.| Market: | Environmental |
| Keywords: | ECD, electron capture detector, environmental, EPA 8081, Organochlorine Pesticides |
| Matrix: | Soil, Water extracts |
| Author: | Andrea Caruso, Massimo Santoro |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.