Find methods for your needs
Anions and Select Organic Acids in Cranberry Juice by Capillary IC
Description
Cap AS19_Cranberry Juice This method demonstrates determination of anions and select organic acids in cranberry juice using capillary IC on IonPac AS19 capillary column and the ICS-5000 capillary system| Market: | Food and Beverage |
| Matrix: | Cranberry juice |
For Research Use Only. Not for use in diagnostic procedures.