Find methods for your needs
Improved size exclusion chromatography of polyquaternium-10
Description
The Thermo Scientific LC system is applied for size exclusion chromatography analysis of polyquaternium-10. The separation is performed on a Thermo Scientific Acclaim SEC-1000 HPLC column, with charged aerosol detection.| Market: | Pharma |
| Keywords: | Acclaim SEC-1000, CAD, Charged aerosol detection, HPLC, Polyquaternium-10, Size exclusion chromatography |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.