Find methods for your needs
Rapid analysis of RNA nucleosides using a Thermo Scientific Accucore 150-Amide-HILIC column
Description
The Thermo Scientific UltiMate 3000RS system is applied for the analysis of RNA nucleosides. The separation is performed on a Thermo Scientific Accucore 150-Amide-HILIC column, with UV detection at 265 nm.| Market: | BioPharma |
| Keywords: | Accucore 150-Amide-HILIC, Adenosine, Cytidine, Guanosine, HPLC, Inosine, Nucleosides, RNA, Uridine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.