Find methods for your needs
AN296: Assay of Fuel-Grade Butanol for Total and Potential Sulfate and Total Chloride Per ASTM D7328-07
Description
Described here is a simple ion chromatography (IC) method to determine total and potential sulfate and total chloride in butanol used as a gasoline additive. It is consistent with the procedure described in ASTM D7328-07, which is intended for the analysis of ethanol samples containing 0.55–20.0 mg/L total sulfate, 4.0–20 mg/L potential sulfate, and 0.75–50.0 mg/L chloride, but can also be applied to butanol samples.| Market: | Petrochem, Oil and Gas |
| Keywords: | AS4A-SC, Chloride, Renewable Energy, Sulfate, Biobutanol, Biofuel, ASTM D4806-11A, High Octane Rating, Blended fuel |
| Matrix: | Butanol |
| Author: | Pranathi Perati, Brian DeBorba, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.