Find methods for your needs
Improved separation of vitamins and other ingredients in an energy drink using a Thermo Scientific PolarAdvantage HPLC column Be the first to rate this application
improved separation vitamins other ingredients an energy drink using a thermo scientific polaradvantage hplc column
# Compound Name Compound Class RT (min)
1 Niacinamide Vitamin 5.67
2 Pyridoxine Vitamin 8.47
3 Caffeine Stimulant 9.53
4 Thiamine Vitamin 10.80
5 Riboflavin Vitamin 11.07
6 Sorbic acid Preservative 14.47
7 Benzoic acid Preservative 14.87
Description
The Thermo Scientific HPLC system is applied for the analysis of vitamins and other ingredients in an energy drink. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) column, with UV detection.
Market: Food and Beverage
Keywords: Acclaim PolarAdvantage (PA), Benzoic acid, Caffeine, Energy drink, HPLC, Niacinamide, Pyridoxine, Riboflavin, Sorbic acid, Thiamine, UV detection, Vitamin
Matrix: Energy drink
Uploaded on 2/26/2015.
For Research Use Only. Not for use in diagnostic procedures.