Find methods for your needs
Improved analysis of spectinomycin and lincomycin using a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of spectinomycin and lincomycin. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column, with electrochemical detection.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage (PA), Electrochemical detection, HPLC, Linomycin, Spectinomycin |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.