Find methods for your needs
Rapid analysis of sulfonamide antibiotics using a Thermo Scientific Acclaim 120 C18 RSLC column
Description
The Thermo Scientific UltiMate 3000 RS HPLC system is applied for the determination of sulfonamide antibiotics. The separation is performed on a Thermo Scientific Acclaim 120 C18 RSLC column, with UV detection at 265 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, Sulfachloropyridazine, Sulfadiazine, Sulfadimethoxine, Sulfamerazine, Sulfamethazine, Sulfapyridine, Sulfaquinoxzline, Sulfathiazole, Sulfonamide antibiotics, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.