Find methods for your needs
High resolution analysis of sulfonamide antibiotic residues in milk using a Thermo Scientific Acclaim 120 C18 column
Description
The Thermo Scientific HPLC system is applied for the determination of sulfonamide antibiotic residues in milk. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 265 nm.| Market: | Food and Beverage |
| Keywords: | HPLC, Milk, Sulfachloropyridazine, Sulfadiazine, Sulfadimethoxine, Sulfamerazine, Sulfamethazine, Sulfapyridine, Sulfaquinoxzline, Sulfathiazole, Sulfonamide antibiotics, Theobromine |
| Matrix: | Milk |
For Research Use Only. Not for use in diagnostic procedures.