Find methods for your needs
AU198: Improved Determination of Trace Concentrations of Oxyhalides and Bromide in Drinking Water Using a Hydroxide-Selective Column
Description
This study demonstrates the Thermo Scientific Dionex IonPac AS27 anion-exchange column for determination of trace concentrations of chlorite, bromate, chlorate, and bromide in drinking water samples. The Dionex IonPac AS27 offers several key advantages over the Dionex AS19 column, including the ability to determine trace bromate in the presence of the EDA preservative, good resolution of dichloroacetate (DCA, a surrogate anion) from potentially interfering matrix anions, and improved resolution between bromate and chloride.| Market: | Environmental |
| Keywords: | Bromate, Chlorate, Chlorite, Dichloroacetate, Dionex AERS 500, Dionex IonPac AS27 Column, Disinfection Byproducts, EPA Method 300.1 (B) |
| Matrix: | Drinking water |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.