Find methods for your needs
High resolution analysis of benzene polycarboxylic acids using a Thermo Scientific Acclaim Organic Acid (OA) HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of benzene polycarboxylic acids. The separation is performed on a Thermo Scientific Acclaim Organic Acid (OA) HPLC column, with UV detection at 230 nm.| Market: | Material Science |
| Keywords: | Acclaim Organic Acid, Benzene polycarboxylic acids, HPLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.