Find methods for your needs
Comparison of the separation of tartaric and quinic acids on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column vs a Thermo Scientific Acclaim Organic Acid column
Description
The Thermo Scientific HPLC system is applied for the analysis of tartaric and quinic acids. The separation achieved on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column is compared to the separation achieved on a Thermo Scientific Acclaim Organic Acid column. The unique chemistry of the Thermo Scientific Acclaim Mixed-Mode WAX-1 column cleanly separates these compounds based on different ionizations.| Market: | Food and Beverage |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acclaim Organic Acid, Comparison, HPLC, Quinic acid, Tartaric acid, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.