Find methods for your needs
Rapid analysis of doxorubicin in human serum using Thermo Scientific SOLA SPE and a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of doxorubicin in human serum. The samples are prepared using Thermo Scientific SOLA solid phase extraction (SPE) cartridges, and the separation performed on a Thermo Scientific Hypersil GOLD HPLC column, with fluorescence detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Daunorubicin, Doxorubicin, Fluorescence detection, HPLC, Human serum, Hypersil GOLD |
| Matrix: | Human serum |
For Research Use Only. Not for use in diagnostic procedures.