Find methods for your needs
Improved analysis of lisinopril by HPLC-UV using a Thermo Scientific Accucore RP-MS column
Description
The Thermo Scientific HPLC system is applied for the analysis of lisinopril. The separation is performed on a Thermo Scientific Accucore RP-MS column, with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Accucore RP-MS, HPLC, Impurities, Lisinopril, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.