Find methods for your needs
Fast analysis of fat-soluble vitamin standards using a Thermo Scientific Acclaim 120 C18 column
Description
The Thermo Scientific HPLC system is applied for the analysis of fat-soluble vitamin standards. The separation is performed using a Thermo Scientific Acclaim 120 C18 column, with UV detection at 280 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, All-trans retinol, Cholecalciferol, Ergocalciferol, Fat-soluble vitamins, HPLC, Tocopherol, UV detection, Vitamin A, Vitamin D2, Vitamin D3, Vitamin E |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.