Find methods for your needs
Fast analysis of loratidine and pseudo-ephedrine in a time-release tablet using a Thermo Scientific Acclaim PolarAdvantage column
Description
The Thermo Scientific HPLC system is applied for the analysis of loratidine and pseudo-ephedrine in a time-release tablet. The separation is performed using a Thermo Scientific Acclaim PolarAdvantage column, with UV detection at 206, 240 and 257 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage, HPLC, Loratidine, Pseudo-ephedrine, Tablet, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.