Find methods for your needs
Evaluation of temperature influence on retention of glycans in HILIC
Description
The Thermo Scientific Dionex Ultimate 3000 BioRS system in combination with the Thermo Scientific Accucore 150-Amide-HILIC is used to run separation of 2-AB-labelled dextran ladder in gradient mode. The column compartment was set at different temperature between 20 and 50 °C. At high temperature the selectivity between oligomers was improved, and peaks were in average narrower. Consequently higher peak capacity was achieved at high temperature.| Market: | Other |
| Keywords: | 2-aminobenzamide, Accucore 150-Amide-HILIC, Biotherapeutics, Fluorescence detection |
| Matrix: | 9/1 (v/v) acetonitrile/ammonium formate pH 4.5 |
For Research Use Only. Not for use in diagnostic procedures.