Find methods for your needs
Fast analysis of clotrimazole in athlete’s foot ointment using a Thermo Scientific Acclaim 120 C8 column
Description
The Thermo Scientific HPLC system is applied for the analysis of clotrimazole in athlete’s foot ointment. The separation is performed using a Thermo Scientific Acclaim 120 C8 column, with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C8, Athlete's foot, Clotrimazole, HPLC, Ointment, UV detection |
| Matrix: | Ointment |
For Research Use Only. Not for use in diagnostic procedures.