Find methods for your needs
AN149: Determination of Chlorite, Bromate, Bromide, and Chlorate in Drinking Water by Ion Chromatography with an On-Line-Generated Postcolumn Reagent.
Description
This application note describes an improved ion chromatography (IC) method to quantify oxyhalide DBP anions and bromide at low concentration levels in reagent water, bottled water, and finished drinking water using an approach that is technically equivalent to U.S. EPA Method 326.0. The oxyhalide anions chlorite, chlorate, bromide, and bromate are separated on a Thermo Scientific Dionex IonPac AS9-HC column and measured by using suppressed conductivity detection (as in EPA Method 300.1), followed by postcolumn reaction (PCR) to enhance detection of bromate.| Market: | Environmental |
| Keywords: | Bottled water, Bromate, Bromide, Chlorate, Chlorite, Drinking Water, EPA, EPA Method 326, External Water Mode, IonPac AS9-HC, Oxyhalides, PC10 reagent delivery, Postcolumn reagent, Suppressed Conductivity Detection |
| Matrix: | Drinking water |
| Author: | David H. Thomas and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.