Find methods for your needs
Simultaneous analysis of multiple counterions using the Acclaim Trinity P2
(1)
Description
The method is based on the Acclaim Trinity P2 column technology. By applying a broad ionic strength gradient over a time period of 11 minutes multiple counterions can be separated simultaneously.
Market: Pharma
Keywords: Charged aerosol detection, Corona Veo, counter ion, Counterion, ion, IonCount
Matrix: N/A
Uploaded on 2/5/2015.
For Research Use Only. Not for use in diagnostic procedures.