Find methods for your needs
IonCount solution reference test for Acclaim Trinity P1 isocratic elutions Be the first to rate this application
Description
The Thermo Scientific™ IonCount Solution is a solution for the analysis of a wide range of pharmaceutical relevant counterions and active pharmaceutical ingredients. The solution is based on a combination of a dedicated column technology and the unique charged aerosol detection. IonCount includes a complete Thermo Scientific™ Dionex™ UltiMate™ 3000 Standard system and a chromatography starter kit. This kit includes two different analytical columns providing alternative selectivity, all required buffer salts for the mobile phases as well as a reference test mix.
Market: Pharma
Keywords: Charged aerosol detection, Corona Veo, counter ion, Counterion, ion, IonCount
Matrix: N/A
Uploaded on 2/5/2015.
For Research Use Only. Not for use in diagnostic procedures.