Find methods for your needs
AN171: Determination of DBP Anions and Bromide in Drinking Water Using a RFIC System with Postcolumn Addition Using EPA 326.0 for Trace Bromate Analysis
Description
In this application, we demonstrate the performance of the Dionex IonPac AS19 column for EPA Method 326.0. This method allows quantification of bromate to 1 μg/L by suppressed conductivity detection with a hydroxide eluent and 0.5 μg/L using postcolumn reaction with UV detection.| Market: | Environmental |
| Keywords: | Bottled Drinking Waters, Bromate, Disinfection Byproduct Anions, Environmental Waters, Thermo Scientific Dionex IonPac AS19 Column |
| Matrix: | Bottled drinking water, Tap water |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.