Find methods for your needs
AN169: Rapid Determination of Phosphate and Citrate in Carbonated Soft Drinks Using a Reagent-Free Ion Chromatography System
Description
This application presents a method using a RFIC system for the simultaneous determination of phosphate and citrate in carbonated soft drinks using the hydroxide-selective Dionex IonPac Fast Anion III column.| Market: | Food and Beverage |
| Keywords: | beverage, Citric Acid, Phosphoric Acid, Rapid Separation, RFIC, Thermo Scientific Dionex Fast Anions III Column, Cola, acidulant |
| Matrix: | Carbonated beverage |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.