Find methods for your needs
AN168: Determination of Trace Concentrations of DBP Anions and Bromide in Drinking Water Using RFIC Followed by Postcolumn Addition of ODA for Trace Bromate Analysis Be the first to rate this application
determination bromate drinking water
Description
In this application note, we demonstrate the performance of the Thermo Scientific Dionex IonPac AS19 column for U.S. EPA Method 317.0. This method combines the advantages of a hydroxide eluent using suppressed conductivity detection with postcolumn addition to further improve the quantification of sub-μg/L bromate.
Market: Environmental
Keywords: Bottled Drinking Waters, Bromate, Disinfection Byproduct Anions, Environmental Waters, EPA 317, Thermo Scientific Dionex IonPac AS19 Column
Matrix: Bottled drinking water, Tap water
Author: Brian De Borba and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 1/29/2015.
For Research Use Only. Not for use in diagnostic procedures.