Find methods for your needs
High resolution separation of cyclophosphamide from polyethylene glycol (PEG) in formulation using a reversed-phase HPLC-UV method
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the determination of cyclophosphamide in formulation. The high resolution separation of cyclophosphamide from PEG was performed on a Thermo Scientific Syncronis C18 HPLC column, with a run time of 20 minutes.| Market: | Pharma |
| Keywords: | Cyclophosphamide, HPLC, Polyethylene glycol (PEG), Syncronis C18, UV detection, Polymer |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.