Find methods for your needs
Accurate determination of l-methyltetrahydrofolate calcium in formulation using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the analysis of a l-methyltetrahydrofolate in solid dosage formulations. The separation is performed on a Thermo Scientific Accucore C8 HPLC column in less than 20 minutes, with UV detection at 280 nm.| Market: | Pharma |
| Keywords: | Accucore C8, Formulation, HPLC, L-methyltetrahydrofolate, UV detection |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.