Find methods for your needs
An accurate determination of mefenamic acid and impurities from formulation using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the analysis of a mefenamic acid in formulation. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column in less than 30 minutes, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | HPLC, Impurities, Mefenamic acid, NSAID, UV detection |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.