Find methods for your needs
An accurate determination of hydroxyzine hydrochloride in tablet formulation using HPLC-UV
Description
The assay of hydroxyzine hydrochloride was performed on a Thermo Scientific Dionex UltiMate 3000 system with UV detection at 232 nm. The Thermo Scientific Hypersil GOLD aQ column was used to perform the assay with a run time of 25 minutes.| Market: | Pharma |
| Keywords: | Antihistamine, HPLC, Hydroxyzine hydrochloride, Hypersil GOLD aQ, UV detection |
| Matrix: | Tablets |
For Research Use Only. Not for use in diagnostic procedures.