Find methods for your needs
An accurate related substances analysis of dexlansoprazole in formulation using reversed-phase HPLC-UV Be the first to rate this application
Description
In this work, the acid degradation products present in the formulation of dexlansoprazole were accurately quantified using the Thermo Scientific Dionex Ultimate 3000 system with UV detection at 285 nm. The separation was performed using a Thermo Scientitic Accucore C18 column with a run time of 25 minutes.
Market: Pharma
Keywords: Accucore C18, Dexlansoprazole, HPLC, Impurities, UV detection
Matrix: Formulation
Uploaded on 12/17/2014.
For Research Use Only. Not for use in diagnostic procedures.