Find methods for your needs
Fast analysis of antibiotics in aqueous environmental matrices using a Thermo Scientific Acclaim RSLC C18 column
Description
The Thermo Scientific HPLC system is applied for the analysis of antibiotics in aqueous environmental matrices. The separation is performed on a Thermo Scientific Acclaim RSLC C18 column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, Antibiotics, Cefamandole, Cefazolin, Cefotaxime, Ceftazidime, Ciprofloxacin, HPLC, Norfloxacin, Penicillin G, Sulfamethazine, UV detection |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.