Find methods for your needs
AN154: Determination of Inorganic Anions in Environmental Waters Using a Hydroxide-Selective Column
Description
In this application, we describe the use of automated eluent generation, combined with a high-capacity, hydroxide-selective, anion-exchange column—the Thermo Scientific Dionex IonPac AS18—for the determination of inorganic anions in environmental waters. The hydroxide-selective Dionex IonPac AS18 column is a superior alternative to the Thermo Scientific Dionex IonPac AS9-HC column using carbonate eluent, which is described in EPA Method 300.1.| Market: | Environmental |
| Keywords: | Common Anions, EPA Method 300.1(A), Hydroxide-Selective, Thermo Scientific Dionex IonPac AS18 Column |
| Matrix: | Environmental water |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.