Find methods for your needs
High resolution separation of lysozyme protein digest using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
Mapping tryptic protein digests can produce highly complex chromatograms where every peak counts. The Thermo Scientific Acclaim 3 μm 2.1 x 100 mm columns yield superior peak capacity and resolution. The 120 C18 chemistry has low silanol activity and high bonding density that produce sharp peaks for small, basic peptides.| Market: | BioPharma |
| Keywords: | Acclaim 120 C18, HPLC, Lysozyme, UV detection, Protein digest |
| Matrix: | Lysozyme digest |
For Research Use Only. Not for use in diagnostic procedures.