Find methods for your needs
An improved separation of Veratrole alcohol and related substances using a Thermo Scientific Accucore PFP HPLC column
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the related substance analysis of veratrole alcohol. The separation of veratrole alcohol and the five related substances was performed on a Thermo Scientific Accucore PFP column with a run time of less than 20 minutes.| Market: | Pharma |
| Keywords: | 3-Methoxyphenol, Accucore PFP, HPLC, MEP, O-methoxyphenol, Phenol, P-hydroxyanisol, UV detection, Veratrole alcohol |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.