Find methods for your needs
An improved separation of Ofloxacin, Ciprofloxacin and impurities using a Thermo Scientific Acclaim 120 C18 HPLC column Be the first to rate this application
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the separation of ofloxacin, ciprofloxacin and impurities. This was achieved using a Thermo Scientific Acclaim 120 C18 column with UV detection at 294 nm.
Market: Pharma
Keywords: Acclaim 120 C18, Antibiotic, Ciprofloxacin, HPLC, Ofloxacin, UV detection
Matrix: N/A
Uploaded on 12/3/2014.
For Research Use Only. Not for use in diagnostic procedures.