Find methods for your needs
AN239: Determination of Iodide in Seawater and Other Saline Matrices Using a Reagent-Free Ion Chromatography System with Suppressed Conductivity and UV Detections
Description
In this application note (AN), a Thermo Scientific™ Dionex™ RFIC-EG™ system with UV detection is used to determine iodide in seawater, synthetic sea salt, and iodized table salt. Iodide is separated from other matrix anions on the Thermo Scientific™ Dionex™ IonPac™ AS20 column, which is a high-capacity column designed for the separation of polarizable anions. The unique selectivity of this column makes it ideal for separating hydrophobic anions in matrices with high concentrations of chloride, sulfate, and carbonate.| Market: | Environmental |
| Keywords: | IC, iodide, saline, Seawater, Suppressed conductivity, UV detection |
| Matrix: | Seawater |
For Research Use Only. Not for use in diagnostic procedures.