Find methods for your needs
A rapid assay and related substances analysis for formulated Vitamin D3 using a Thermo Scientific Accucore XL C18 HPLC column
Description
The Thermo Scientific Ultimate 3000 RS system is applied for the analysis of vitamin D tablets. The separation was performed on the Thermo Scientific Accucore XL C18 column with a run time of 10 minutes.| Market: | Pharma |
| Keywords: | Accucore XL C18, HPLC, UV detection, Vitamin D3 |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.