Find methods for your needs
Ethinylestradiol tablets assay according to the Chinese Pharmacopoeia method using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of ethinylestradiol tablets according to the Chinese Pharmacopoeia method. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Chinese pharmacopeia, Ethinylestradiol, HPLC, Tablet, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.