Find methods for your needs
Rapid separation of five herbicides using a Thermo Scientific Acclaim 120 C18 HPLC column Be the first to rate this application
rapid separation five herbicides using a thermo scientific acclaim 120 c18 hplc column
# Compound Name Compound Class RT (min)
1 Monuron Herbicide 1.90
2 Atrazine Herbicide 2.50
3 Diuron Herbicide 2.80
4 Propazine Herbicide 3.50
5 Linuron Herbicide 3.60
Description
The fast separation of five herbicides was performed using the Thermo Scientific Acclaim 120 C18 column with a run time of less than five minutes with MS detection.
Market: Environmental
Keywords: Acclaim 120 C18, Atrazine, Diuron, Herbicides, Linuron, Monuron, MS detection, Propazine
Matrix: N/A
Uploaded on 11/10/2014.
For Research Use Only. Not for use in diagnostic procedures.